Imprimer

Anti-CD41 gpIIb IIIa Mouse Monoclonal Antibody (FITC (Fluorescein))

Fournisseur: Biorbyt
BIRBORB26532-100EA 0 CHF
BIRBORB26532-100
Anti-CD41 gpIIb IIIa Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorps
Flow Cytometry: 20 ?l/5 x105

Type: Primary
Antigen: CD41 gpIIb IIIa
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR