Imprimer

Anti-ITGB1 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26525-0.2EA 944 CHF
BIRBORB26525-0.2
Anti-ITGB1 Mouse Monoclonal Antibody
Anticorps
Flow Cytometry: 2 ?g/5 x105

Type: Primary
Antigen: ITGB1 (integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR