Imprimer

Anti-CEACAM8 Mouse Monoclonal Antibody (FITC (Fluorescein))

Fournisseur: Biorbyt
BIRBORB26519-100EA 0 CHF
BIRBORB26519-100
Anti-CEACAM8 Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorps
Flow Cytometry: 20 ?l/106

Type: Primary
Antigen: CEACAM8 (carcinoembryonic antigen-related cell adhesion molecule 8)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR