Imprimer

Anti-ITGAM Mouse Monoclonal Antibody (FITC (Fluorescein))

Fournisseur: Biorbyt
BIRBORB26518-100EA 0 CHF
BIRBORB26518-100
Anti-ITGAM Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticorps
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR