Imprimer

Anti-CD77 Rat Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26497-100EA 0 CHF
BIRBORB26497-100
Anti-CD77 Rat Monoclonal Antibody
Anticorps
Immunofluorescence: 20 ?l for5x105

Type: Primary
Antigen: CD77
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgM Kappa (Rat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR