Imprimer

Anti-Mouse CD71 Rat Monoclonal Antibody (PE (Phycoerythrin))

Fournisseur: Biorbyt
BIRBORB26429-100EA 0 CHF
BIRBORB26429-100
Anti-Mouse CD71 Rat Monoclonal Antibody (PE (Phycoerythrin))
Anticorps
Type: Primary
Antigen: Mouse CD71
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a (Rat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR