Imprimer

Anti-Mouse CD4 (L3T4) Rat Monoclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB26380-0.5EA 727 CHF
BIRBORB26380-0.5
Anti-Mouse CD4 (L3T4) Rat Monoclonal Antibody (Biotin)
Anticorps
Flow Cytometry: 1 ?g

Type: Primary
Antigen: Mouse CD4 (L3T4)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b (Rat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR