Imprimer

Anti-CD56 NCAM Mouse Monoclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB26366-0.2EA 0 CHF
BIRBORB26366-0.2
Anti-CD56 NCAM Mouse Monoclonal Antibody (Biotin)
Anticorps
Flow Cytometry: 1 ?g

Type: Primary
Antigen: CD56 NCAM
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR