Imprimer

Anti-Factor XI Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26328-100EA 0 CHF
BIRBORB26328-100
Anti-Factor XI Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Factor XI
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR