Imprimer

Anti-F2R Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB26327-0.2EA 0 CHF
BIRBORB26327-0.2
Anti-F2R Mouse monoclonal antibody
Anticorps
Flow Cytometry: 2 ?g

Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR