Imprimer

Anti-D-dimer Mouse Monoclonal Antibody (Biotin)

Fournisseur: Biorbyt
BIRBORB26322-1EA 1753 CHF
BIRBORB26322-1
Anti-D-dimer Mouse Monoclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: D-dimer
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR