Imprimer

Anti-D-dimer Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB26316-1EA 758 CHF
BIRBORB26316-1
Anti-D-dimer Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: D-dimer
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR