Imprimer

Anti-F8 Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB26280-1EA 0 CHF
BIRBORB26280-1
Anti-F8 Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: F8 (coagulation factor VIII, procoagulant component)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR