Imprimer

Anti-CD97 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26131-100EA 424 CHF
BIRBORB26131-100
Anti-CD97 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: CD97 (CD97 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR