Imprimer

Anti-CD300A Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26112-100EA 476 CHF
BIRBORB26112-100
Anti-CD300A Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: CD300A (CD300a molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR