Imprimer

Anti-FCGR3A Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB26105-100EA 476 CHF
BIRBORB26105-100
Anti-FCGR3A Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR