Imprimer

Anti-FSH Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26064-1EA 626 CHF
BIRBORB26064-1
Anti-FSH Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: FSH
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR