Imprimer

Anti-FSH Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26061-1EA 607 CHF
BIRBORB26061-1
Anti-FSH Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: FSH
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR