Imprimer

Anti-Tobramycin Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26060-1EA 566 CHF
BIRBORB26060-1
Anti-Tobramycin Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Tobramycin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR