Imprimer

Anti-MYC Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB26044-0.2EA 607 CHF
BIRBORB26044-0.2
Anti-MYC Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR