Imprimer

Anti-Alpha Fetoprotein Mouse Monoclonal Antibody (Biotin) [clone: 4A3]

Fournisseur: Biorbyt
BIRBORB26008-1EA 1214 CHF
BIRBORB26008-1
Anti-Alpha Fetoprotein Mouse Monoclonal Antibody (Biotin) [clone: 4A3]
Anticorps
Type: Primary
Antigen: Alpha Fetoprotein
Clonality: Monoclonal
Clone: 4A3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR