Imprimer

Anti-PSE1/Ku Protein Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB25973-0.2EA 918 CHF
BIRBORB25973-0.2
Anti-PSE1/Ku Protein Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PSE1/Ku Protein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR