Imprimer

Anti-Laminin 5 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB25925-100EA 0 CHF
BIRBORB25925-100
Anti-Laminin 5 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Laminin 5
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR