Imprimer

Anti-FOLH1 Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB25888-1EA 0 CHF
BIRBORB25888-1
Anti-FOLH1 Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: FOLH1 (folate hydrolase (prostate-specific membrane antigen) 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR