Imprimer

Anti-Lambda Light Chain Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25810-10EA 539 CHF
BIRBORB25810-10
Anti-Lambda Light Chain Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Lambda Light Chain
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR