Imprimer

Anti-C4A Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25808-10EA 462 CHF
BIRBORB25808-10
Anti-C4A Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: C4A (complement component 4A (Rodgers blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR