Imprimer

Anti-APOC3 Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25779-1EA 1122 CHF
BIRBORB25779-1
Anti-APOC3 Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:200-1:40000

Type: Primary
Antigen: APOC3 (apolipoprotein C-III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR