Imprimer

Anti-Apo (a) Sheep Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25777-1EA 895 CHF
BIRBORB25777-1
Anti-Apo (a) Sheep Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Apo (a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR