Imprimer

Anti-AFP Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25752-1EA 521 CHF
BIRBORB25752-1
Anti-AFP Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR