Imprimer

Anti-PSA, Total Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25751-1EA 399 CHF
BIRBORB25751-1
Anti-PSA, Total Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: PSA, Total
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR