Imprimer

Anti-Superoxide Dismutase Sheep Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25737-1EA 926 CHF
BIRBORB25737-1
Anti-Superoxide Dismutase Sheep Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Superoxide Dismutase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR