Imprimer

Anti-Fluorescein Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB25583-1EA 755 CHF
BIRBORB25583-1
Anti-Fluorescein Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: Fluorescein (fluorescein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR