Imprimer

Anti-Aflatoxin Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB25573-1EA 835 CHF
BIRBORB25573-1
Anti-Aflatoxin Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Aflatoxin (aflatoxin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR