Imprimer

Anti-CTNNB1 Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB25512-100EA 0 CHF
BIRBORB25512-100
Anti-CTNNB1 Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: CTNNB1 (catenin (cadherin-associated protein), beta 1, 88kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR