Imprimer

Anti-Fluorescein Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25487-1EA 667 CHF
BIRBORB25487-1
Anti-Fluorescein Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Fluorescein (fluorescein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR