Imprimer

Anti-PLAT Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25483-0.2EA 754 CHF
BIRBORB25483-0.2
Anti-PLAT Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:10000-1:100000; Western Blot: 1:1000-1:10000; Immunochemistry: 1:100-1:1000

Type: Primary
Antigen: PLAT (plasminogen activator, tissue)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR