Imprimer

Anti-ADH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Biorbyt
BIRBORB25409-20EA 1112 CHF
BIRBORB25409-20
Anti-ADH Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
ELISA: 1:20000-1:100000

Type: Primary
Antigen: ADH (alcohol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR