Imprimer
Anti-PEP Carboxylase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Fournisseur: Biorbyt
BIRBORB25408-20EA 911 CHF
BIRBORB25408-20
Anti-PEP Carboxylase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
ELISA: 1:1000-1:6000
Type: Primary
Antigen: PEP Carboxylase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now