Imprimer

Anti-Rat ANP Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25299-100EA 0 CHF
BIRBORB25299-100
Anti-Rat ANP Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Rat ANP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR