Imprimer

Anti-BNP Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25296-100EA 0 CHF
BIRBORB25296-100
Anti-BNP Rabbit Polyclonal Antibody
Anticorps
Immunofluorescence: 1:1000

Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR