Imprimer

Anti-CCND2 Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB25268-5EA 0 CHF
BIRBORB25268-5
Anti-CCND2 Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: CCND2 (cyclin D2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR