Imprimer

Anti-Oxidized Lp(a) Rabbit Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25196-20EA 0 CHF
BIRBORB25196-20
Anti-Oxidized Lp(a) Rabbit Polyclonal Antibody
Anticorps
ELISA: 1:500-1:5000

Type: Primary
Antigen: Oxidized Lp(a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR