Imprimer

Anti-Ribosomal P Antigen Human Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25155-2EA 518 CHF
BIRBORB25155-2
Anti-Ribosomal P Antigen Human Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Ribosomal P Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Human
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR