Imprimer

Anti-APOB Goat Polyclonal Antibody

Fournisseur: Biorbyt
BIRBORB25015-10EA 492 CHF
BIRBORB25015-10
Anti-APOB Goat Polyclonal Antibody
Anticorps
Type: Primary
Antigen: APOB (apolipoprotein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR