Imprimer

Anti-sLOX-1 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24999-1EA 1257 CHF
BIRBORB24999-1
Anti-sLOX-1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: sLOX-1
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR