Imprimer

Anti-TNNT2 Mouse Monoclonal Antibody [clone: 9G6]

Fournisseur: Biorbyt
BIRBORB24989-1EA 871 CHF
BIRBORB24989-1
Anti-TNNT2 Mouse Monoclonal Antibody [clone: 9G6]
Anticorps
Type: Primary
Antigen: TNNT2 (troponin T type 2 (cardiac))
Clonality: monoclonal
Clone: 9G6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR