Imprimer

Anti-TNNI3 Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24982-1EA 1142 CHF
BIRBORB24982-1
Anti-TNNI3 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR