Imprimer

Anti-BNP Mouse monoclonal antibody

Fournisseur: Biorbyt
BIRBORB24966-1EA 1382 CHF
BIRBORB24966-1
Anti-BNP Mouse monoclonal antibody
Anticorps
Type: Primary
Antigen: BNP (Brain Natriuretic Peptide)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR