Imprimer

Anti-Troponin I, Skeletal Muscle Mouse Monoclonal Antibody

Fournisseur: Biorbyt
BIRBORB24851-1EA 1040 CHF
BIRBORB24851-1
Anti-Troponin I, Skeletal Muscle Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: Troponin I, Skeletal Muscle
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR