Imprimer

Anti-HB Mouse monoclonal antibody [clone: 7A4]

Fournisseur: Biorbyt
BIRBORB24755-1EA 790 CHF
BIRBORB24755-1
Anti-HB Mouse monoclonal antibody [clone: 7A4]
Anticorps
Type: Primary
Antigen: HB (hemoglobin)
Clonality: monoclonal
Clone: 7A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR